Assay Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568873.2 | XP_005568930.1 | 1 - 2 | 234 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005568873.2 transcripts.
Set Membership: |
|

ROMO1
Genomic Map