Assay Details
| Gene Symbol: CRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562413.2 | XP_005562470.1 | 1 - 2 | 160 | 91 | |
| XM_005562415.2 | XP_005562472.1 | 1 - 2 | 160 | 91 | ||
| XM_015454280.1 | XP_015309766.1 | 1 - 2 | 160 | 91 | ||
More Information
Set Membership: |
|

CRIP2
Genomic Map