Assay Details
| Gene Symbol: FCGR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284040.1 | NP_001270969.1 | 4 - 5 | 586 | 78 | |
| GenBank mRNA | AB170928.1 | - | 4 - 5 | 586 | 78 | |
| AF485812.1 | - | 4 - 5 | 551 | 78 | ||
More Information
Set Membership: |
|

FCGR1A
Genomic Map