Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285215.1 | NP_001272144.1 | 1 - 2 | 241 | 99 | |
| GenBank mRNA | AB171570.1 | - | 1 - 2 | 241 | 99 | |
More Information
Set Membership: |
|

SNAI2
Genomic Map