Assay Details
| Gene Symbol: LRRC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542788.2 | XP_005542845.1 | 6 - 7 | 2731 | 89 | |
| XM_005542790.2 | XP_005542847.1 | 4 - 5 | 2432 | 89 | ||
| XM_015433112.1 | XP_015288598.1 | 5 - 6 | 2595 | 89 | ||
| XM_015433115.1 | XP_015288601.1 | 5 - 6 | 5771 | 89 | ||
| XM_015433121.1 | XP_015288607.1 | 4 - 5 | 2494 | 89 | ||
| XM_015433129.1 | XP_015288615.1 | 5 - 6 | 2595 | 89 | ||
More Information
Set Membership: |
|

LRRC8B
Genomic Map