Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594634.2 | XP_005594691.1 | 6 | 1421 | 104 | |
| XM_005594637.2 | XP_005594694.1 | 5 | 1259 | 104 | ||
More Information
Set Membership: |
|

MOSPD1
Genomic Map