Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283923.1 | NP_001270852.1 | 3 - 4 | 970 | 116 | |
| XM_015440960.1 | XP_015296446.1 | 4 - 5 | 767 | 116 | ||
| XM_015440962.1 | XP_015296448.1 | 4 - 5 | 793 | 116 | ||
| GenBank mRNA | AB060218.1 | - | 3 - 4 | 970 | 116 | |
More Information
Set Membership: |
|

ZFP30
Genomic Map