Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284887.1 | NP_001271816.1 | 4 - 5 | 899 | 85 | |
XM_005582134.2 | XP_005582191.1 | 5 - 6 | 817 | 85 | ||
GenBank mRNA | AB220416.1 | - | 4 - 5 | 899 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
