Assay Details
Gene Symbol: GSKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283559.1 | NP_001270488.1 | 2 - 3 | 317 | 80 | |
XM_005562161.2 | XP_005562218.1 | 2 - 3 | 279 | 80 | ||
XM_005562162.2 | XP_005562219.1 | 2 - 3 | 279 | 80 | ||
XM_015453205.1 | XP_015308691.1 | 2 - 3 | 283 | 80 | ||
GenBank mRNA | AB168649.1 | - | 2 - 3 | 317 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
