Assay Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287660.1 | NP_001274589.1 | 3 - 4 | 334 | 79 | |
| XM_005540088.2 | XP_005540145.1 | 4 - 5 | 430 | 79 | ||
| XM_005540089.2 | XP_005540146.1 | 4 - 5 | 430 | 79 | ||
| XM_005540091.2 | XP_005540148.1 | 4 - 5 | 322 | 79 | ||
| XM_005540092.2 | XP_005540149.1 | 3 - 4 | 258 | 79 | ||
| XM_005540093.2 | XP_005540150.1 | 3 - 4 | 258 | 79 | ||
| XM_015446429.1 | XP_015301915.1 | 4 - 5 | 441 | 79 | ||
| XM_015446557.1 | XP_015302043.1 | 3 - 4 | 364 | 79 | ||
| XM_015446588.1 | XP_015302074.1 | 4 - 5 | 322 | 79 | ||
| XM_015446644.1 | XP_015302130.1 | 3 - 4 | 256 | 79 | ||
| GenBank mRNA | AB168132.1 | - | 3 - 4 | 334 | 79 | |
More Information
Set Membership: |
|

TEX35
Genomic Map