Assay Details
| Gene Symbol: CCDC89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287725.1 | NP_001274654.1 | 1 | 1023 | 62 | |
| XM_015435362.1 | XP_015290848.1 | 1 | 1442 | 62 | ||
| XM_015435363.1 | XP_015290849.1 | 1 | 1442 | 62 | ||
| XM_015435364.1 | XP_015290850.1 | 1 | 1442 | 62 | ||
| XM_015435365.1 | XP_015290851.1 | 1 | 1442 | 62 | ||
| XM_015435366.1 | XP_015290852.1 | 1 | 1442 | 62 | ||
| XM_015435367.1 | XP_015290853.1 | 1 | 1442 | 62 | ||
| XM_015435368.1 | XP_015290854.1 | 1 | 1442 | 62 | ||
| XM_015435369.1 | XP_015290855.1 | 1 | 1442 | 62 | ||
| XM_015435370.1 | XP_015290856.1 | 1 | 1442 | 62 | ||
| XM_015435371.1 | XP_015290857.1 | 1 | 1442 | 62 | ||
| XM_015435372.1 | XP_015290858.1 | 1 | 1442 | 62 | ||
| GenBank mRNA | AB070192.1 | - | 1 | 1023 | 62 | |
More Information
Set Membership: |
|

CCDC89
Genomic Map