Assay Details
Gene Symbol: RQCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283775.1 | NP_001270704.1 | 8 | 2193 | 75 | |
XM_005574286.2 | XP_005574343.1 | 8 | 3061 | 93 | ||
XM_015432080.1 | XP_015287566.1 | 7 | 2391 | 93 | ||
XM_015432081.1 | XP_015287567.1 | 7 | 2977 | 93 | ||
XM_015432083.1 | XP_015287569.1 | 8 | 2239 | 93 | ||
GenBank mRNA | AB179421.1 | - | 8 | 2193 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
