Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284620.1 | NP_001271549.1 | 2 - 3 | 435 | 83 | |
| XM_005540871.2 | XP_005540928.1 | 2 - 3 | 450 | 83 | ||
| GenBank mRNA | AB174552.1 | - | 2 - 3 | 435 | 83 | |
More Information
Set Membership: |
|

RRP15
Genomic Map