Assay Details
| Gene Symbol: SIAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284617.1 | NP_001271546.1 | 3 - 4 | 436 | 73 | |
| GenBank mRNA | AB170267.1 | - | 3 - 4 | 436 | 73 | |
More Information
Set Membership: |
|

SIAE
Genomic Map