Assay Details
| Gene Symbol: TCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283913.1 | NP_001270842.1 | 1 - 2 | 1385 | 98 | |
| GenBank mRNA | AB172759.1 | - | 1 - 2 | 1385 | 98 | |
More Information
Set Membership: |
|

TCAP
Genomic Map