Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284610.1 | NP_001271539.1 | 4 - 5 | 644 | 76 | |
| XM_015440947.1 | XP_015296433.1 | 4 - 5 | 750 | 76 | ||
| XM_015440948.1 | XP_015296434.1 | 4 - 5 | 541 | 76 | ||
| XM_015440949.1 | XP_015296435.1 | 4 - 5 | 479 | 76 | ||
| GenBank mRNA | AB178992.1 | - | 4 - 5 | 646 | 76 | |
More Information
Set Membership: |
|

ZNF432
Genomic Map