Assay Details
Gene Symbol: LOC101926300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284618.1 | NP_001271547.1 | 1 | 921 | 69 | |
GenBank mRNA | AB046010.1 | - | 1 | 921 | 69 |
Gene Symbol: ATXN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442521.1 | XP_015298007.1 | 1 | 272 | 69 | |
XM_015442524.1 | XP_015298010.1 | 1 | 272 | 69 | ||
XM_015442526.1 | XP_015298012.1 | 1 | 272 | 69 | ||
XM_015442527.1 | XP_015298013.1 | 1 | 272 | 69 | ||
XM_015442528.1 | XP_015298014.1 | 1 | 272 | 69 | ||
XM_015442529.1 | XP_015298015.1 | 1 | 272 | 69 | ||
XM_015442531.1 | XP_015298017.1 | 1 | 271 | 69 | ||
XM_015442532.1 | XP_015298018.1 | 1 | 271 | 69 | ||
XM_015442533.1 | XP_015298019.1 | 1 | 271 | 69 | ||
XM_015442534.1 | XP_015298020.1 | 1 | 271 | 69 | ||
XM_015442535.1 | XP_015298021.1 | 1 | 271 | 69 | ||
XM_015442536.1 | XP_015298022.1 | 1 | 271 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015442521.1, XM_015442524.1, XM_015442526.1, XM_015442527.1, XM_015442528.1, XM_015442529.1, XM_015442531.1, XM_015442532.1, XM_015442533.1, XM_015442534.1, XM_015442535.1, XM_015442536.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
