Assay Details
| Gene Symbol: SRSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285072.1 | NP_001272001.1 | 1 - 2 | 534 | 87 | |
| XM_005585084.2 | XP_005585141.1 | 1 - 2 | 598 | 87 | ||
| GenBank mRNA | AB168918.1 | - | 1 - 2 | 536 | 87 | |
| AB170761.1 | - | 1 - 2 | 534 | 87 | ||
| AB170941.1 | - | 1 - 2 | 536 | 87 | ||
More Information
Set Membership: |
|

SRSF2
Genomic Map