Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285824.1 | NP_001272753.1 | 3 - 4 | 659 | 61 | |
| GenBank mRNA | DQ078721.1 | - | 3 - 4 | 659 | 61 | |
| DQ529984.1 | - | 1 - 2 | 37 | 61 | ||
More Information
Set Membership: |
|

HSD17B2
Genomic Map