Assay Details
| Gene Symbol: NDUFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287185.1 | NP_001274114.1 | 1 - 2 | 166 | 102 | |
| GenBank mRNA | AB072016.1 | - | 1 - 2 | 166 | 102 | |
| JQ794781.1 | - | 1 - 2 | 166 | 102 | ||
More Information
Set Membership: |
|

NDUFC2
Genomic Map