Assay Details
| Gene Symbol: LPAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284891.1 | NP_001271820.1 | 5 | 1324 | 99 | |
| XM_005581086.2 | XP_005581143.1 | 5 | 1600 | 99 | ||
| XM_005581087.2 | XP_005581144.1 | 5 | 1666 | 99 | ||
| XM_005581088.2 | XP_005581145.1 | 5 | 2927 | 99 | ||
| XM_005581089.2 | XP_005581146.1 | 5 | 1487 | 99 | ||
| GenBank mRNA | AB172806.1 | - | 5 | 1324 | 99 | |
More Information
Set Membership: |
|

LPAR1
Genomic Map