Assay Details
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545248.2 | XP_005545305.1 | 1 - 2 | 320 | 81 | |
XM_015444842.1 | XP_015300328.1 | 1 - 2 | 320 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
