Assay Details
| Gene Symbol: CALHM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551647.2 | XP_005551704.1 | 1 - 2 | 736 | 94 | |
| XM_015448584.1 | XP_015304070.1 | 2 - 3 | 251 | 94 | ||
More Information
Set Membership: |
|

CALHM4
Genomic Map