Assay Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439538.1 | XP_015295024.1 | 5 - 6 | 563 | 103 | |
| XM_015439539.1 | XP_015295025.1 | 5 - 6 | 551 | 103 | ||
| XM_015439540.1 | XP_015295026.1 | 5 - 6 | 517 | 103 | ||
| XM_015439541.1 | XP_015295027.1 | 5 - 6 | 476 | 103 | ||
| XM_015439542.1 | XP_015295028.1 | 4 - 5 | 445 | 103 | ||
| XM_015439543.1 | XP_015295029.1 | 4 - 5 | 398 | 103 | ||
| GenBank mRNA | AB168972.1 | - | 5 - 6 | 3498 | 103 | |
More Information
Set Membership: |
|

SPACA7
Genomic Map