Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589911.2 | XP_005589968.1 | 6 - 7 | 1120 | 64 | |
XM_005589912.2 | XP_005589969.1 | 6 - 7 | 1121 | 64 | ||
XM_005589914.2 | XP_005589971.1 | 6 - 7 | 1137 | 64 | ||
XM_015441138.1 | XP_015296624.1 | 5 - 6 | 970 | 64 | ||
XM_015441139.1 | XP_015296625.1 | 5 - 6 | 969 | 64 | ||
XM_015441140.1 | XP_015296626.1 | 5 - 6 | 970 | 64 | ||
XM_015441141.1 | XP_015296627.1 | 5 - 6 | 924 | 64 | ||
XM_015441142.1 | XP_015296628.1 | 5 - 6 | 922 | 64 | ||
XM_015441143.1 | XP_015296629.1 | 5 - 6 | 924 | 64 | ||
XM_015441144.1 | XP_015296630.1 | 4 - 5 | 917 | 64 | ||
XM_015441145.1 | XP_015296631.1 | 5 - 6 | 778 | 64 | ||
XM_015441146.1 | XP_015296632.1 | 5 - 6 | 777 | 64 | ||
XM_015441147.1 | XP_015296633.1 | 5 - 6 | 778 | 64 | ||
XM_015441148.1 | XP_015296634.1 | 6 - 7 | 1308 | 64 | ||
XM_015441149.1 | XP_015296635.1 | 6 - 7 | 1121 | 64 | ||
XM_015441150.1 | XP_015296636.1 | 6 - 7 | 1070 | 64 | ||
XM_015441151.1 | XP_015296637.1 | 5 - 6 | 549 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
