Assay Details
| Gene Symbol: LOC102124147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590008.2 | XP_005590065.1 | 3 - 4 | 162 | 113 | |
| XM_015441358.1 | XP_015296844.1 | 27 - 28 | 3499 | 113 | ||
| XM_015441359.1 | XP_015296845.1 | 1 - 2 | 66 | 113 | ||
More Information
Set Membership: |
|

LOC102124147
Genomic Map