Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569730.2 | XP_005569787.1 | 2 - 3 | 675 | 96 | |
| XM_005569731.2 | XP_005569788.1 | 2 - 3 | 599 | 96 | ||
| XM_015430209.1 | XP_015285695.1 | 2 - 3 | 599 | 96 | ||
| XM_015430210.1 | XP_015285696.1 | 2 - 3 | 518 | 96 | ||
| XM_015430211.1 | XP_015285697.1 | 2 - 3 | 727 | 96 | ||
| XM_015430212.1 | XP_015285698.1 | 2 - 3 | 325 | 96 | ||
More Information
Set Membership: |
|

NINJ2
Genomic Map