Assay Details
| Gene Symbol: PYCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585284.2 | XP_005585341.1 | 6 - 7 | 899 | 91 | |
| XM_005585287.2 | XP_005585344.1 | 6 - 7 | 737 | 91 | ||
| XM_015437957.1 | XP_015293443.1 | 4 - 5 | 951 | 91 | ||
| XM_015437958.1 | XP_015293444.1 | 5 - 6 | 829 | 91 | ||
| XM_015437959.1 | XP_015293445.1 | 4 - 5 | 842 | 91 | ||
More Information
Set Membership: |
|

PYCR1
Genomic Map