Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588007.1 | XP_005588064.1 | 1 - 2 | 154 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANGPTL8| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588007.1 | XP_005588064.1 | 1 - 2 | 154 | 100 | |