Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588007.1 | XP_005588064.1 | 4 - 5 | 735 | 49 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANGPTL8| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588007.1 | XP_005588064.1 | 4 - 5 | 735 | 49 | |
Set Membership: |
|