Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546799.2 | XP_005546856.1 | 2 - 3 | 594 | 72 | |
| XM_005546800.2 | XP_005546857.1 | 2 - 3 | 835 | 72 | ||
| XM_015445717.1 | XP_015301203.1 | 2 - 3 | 597 | 72 | ||
More Information
Set Membership: |
|

ZNF662
Genomic Map