Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546799.2 | XP_005546856.1 | 3 - 4 | 702 | 93 | |
| XM_005546800.2 | XP_005546857.1 | 3 - 4 | 943 | 93 | ||
| XM_015445716.1 | XP_015301202.1 | 3 - 4 | 699 | 93 | ||
| XM_015445718.1 | XP_015301204.1 | 3 - 4 | 1077 | 93 | ||
| XM_015445719.1 | XP_015301205.1 | 3 - 4 | 350 | 93 | ||
| XM_015445720.1 | XP_015301206.1 | 3 - 4 | 705 | 93 | ||
| XM_015445721.1 | XP_015301207.1 | 3 - 4 | 1074 | 93 | ||
More Information
Set Membership: |
|

ZNF662
Genomic Map