Assay Details
Gene Symbol: SEC14L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568097.2 | XP_005568154.1 | 11 - 12 | 1149 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SEC14L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568097.2 | XP_005568154.1 | 11 - 12 | 1149 | 63 |
Set Membership: |
![]() ![]() ![]() ![]() |