Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572519.2 | XP_005572576.1 | 3 - 4 | 1225 | 95 | |
| XM_005572520.2 | XP_005572577.1 | 3 - 4 | 1427 | 95 | ||
| XM_015431700.1 | XP_015287186.1 | 3 - 4 | 1089 | 95 | ||
More Information
Set Membership: |
|

VPS37B
Genomic Map