Assay Details
| Gene Symbol: TGM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568524.1 | XP_005568581.1 | 4 - 5 | 604 | 56 | |
| XM_005568525.1 | XP_005568582.1 | 4 - 5 | 604 | 56 | ||
More Information
Set Membership: |
|

TGM6
Genomic Map