Assay Details
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555868.2 | XP_005555925.1 | 3 - 4 | 546 | 101 | |
| XM_005555870.2 | XP_005555927.1 | 3 - 4 | 815 | 101 | ||
| XM_015450806.1 | XP_015306292.1 | 3 - 4 | 587 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005555870.2, XM_015450806.1 transcripts.
Set Membership: |
|

NUDT6
Genomic Map