Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581922.2 | XP_005581979.1 | 3 - 4 | 1270 | 104 | |
| XM_005581923.2 | XP_005581980.1 | 3 - 4 | 779 | 104 | ||
| XM_015437276.1 | XP_015292762.1 | 3 - 4 | 719 | 104 | ||
More Information
Set Membership: |
|

ABHD17B
Genomic Map