Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587527.2 | XP_005587584.1 | 2 - 3 | 329 | 74 | |
| XM_005587528.2 | XP_005587585.1 | 2 - 3 | 329 | 74 | ||
| XM_005587529.2 | XP_005587586.1 | 2 - 3 | 329 | 74 | ||
More Information
Set Membership: |
|

MRPL54
Genomic Map