Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443112.1 | XP_015298598.1 | 5 - 6 | 666 | 56 | |
| XM_015443113.1 | XP_015298599.1 | 5 - 6 | 606 | 56 | ||
More Information
Set Membership: |
|

OSGIN1
Genomic Map