Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585097.2 | XP_005585154.1 | 10 - 11 | 2023 | 60 | |
XM_005585099.2 | XP_005585156.1 | 10 - 11 | 2186 | 60 | ||
XM_005585100.2 | XP_005585157.1 | 11 - 12 | 1752 | 60 | ||
XM_005585101.1 | XP_005585158.1 | 10 - 11 | 2378 | 60 | ||
XM_005585102.2 | XP_005585159.1 | 9 - 10 | 1168 | 60 | ||
XM_015438892.1 | XP_015294378.1 | 10 - 11 | 1695 | 60 | ||
XM_015438893.1 | XP_015294379.1 | 9 - 10 | 1058 | 60 | ||
XM_015438894.1 | XP_015294380.1 | 9 - 10 | 1271 | 60 | ||
XM_015438895.1 | XP_015294381.1 | 10 - 11 | 1330 | 60 | ||
XM_015438896.1 | XP_015294382.1 | 9 - 10 | 1230 | 60 | ||
XM_015438897.1 | XP_015294383.1 | 9 - 10 | 979 | 60 | ||
XM_015438898.1 | XP_015294384.1 | 9 - 10 | 1134 | 60 | ||
GenBank mRNA | AB179041.1 | - | 7 - 8 | 660 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
