Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591658.2 | XP_005591715.1 | 1 - 2 | 821 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF771| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591658.2 | XP_005591715.1 | 1 - 2 | 821 | 104 | |
Set Membership: |
|