Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591658.2 | XP_005591715.1 | 2 - 3 | 968 | 143 | |
| XM_005591659.2 | XP_005591716.1 | 2 - 3 | 986 | 143 | ||
More Information
Set Membership: |
|

ZNF771
Genomic Map