Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584884.2 | XP_005584941.1 | 4 - 5 | 413 | 105 | |
| XM_005584885.2 | XP_005584942.1 | 4 - 5 | 413 | 105 | ||
More Information
Set Membership: |
|

TMEM104
Genomic Map