Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590147.2 | XP_005590204.1 | 5 - 6 | 697 | 105 | |
| XM_005590148.2 | XP_005590205.1 | 4 - 5 | 379 | 105 | ||
| XM_005590149.2 | XP_005590206.1 | 4 - 5 | 1039 | 105 | ||
| XM_015441333.1 | XP_015296819.1 | 4 - 5 | 1121 | 105 | ||
| XM_015441334.1 | XP_015296820.1 | 3 - 4 | 2397 | 105 | ||
| XM_015441335.1 | XP_015296821.1 | 5 - 6 | 619 | 105 | ||
| XM_015441336.1 | XP_015296822.1 | 4 - 5 | 602 | 105 | ||
| XM_015441338.1 | XP_015296824.1 | 5 - 6 | 692 | 105 | ||
More Information
Set Membership: |
|

ZNF613
Genomic Map