Assay Details
| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554056.2 | XP_005554113.1 | 3 - 4 | 463 | 110 | |
| XM_015449907.1 | XP_015305393.1 | 3 - 4 | 178 | 110 | ||
More Information
Set Membership: |
|

TMEM14C
Genomic Map