Assay Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287281.1 | NP_001274210.1 | 1 - 2 | 196 | 69 | |
| GenBank mRNA | AB072796.1 | - | 1 - 2 | 196 | 69 | |
More Information
Set Membership: |
|

SPESP1
Genomic Map