Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319482.1 | NP_001306411.1 | 4 - 5 | 1159 | 59 | |
| XM_005560089.2 | XP_005560146.1 | 4 - 5 | 1221 | 59 | ||
| GenBank mRNA | D17575.1 | - | 4 - 5 | 1159 | 59 | |
More Information
Set Membership: |
|

CYP1A1
Genomic Map