Assay Details
| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439611.1 | XP_015295097.1 | 1 - 2 | 156 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM21| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439611.1 | XP_015295097.1 | 1 - 2 | 156 | 80 | |