Assay Details
| Gene Symbol: FIGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549590.2 | XP_005549647.1 | 3 - 4 | 293 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005549590.2 transcripts.
Set Membership: |
|

FIGNL1
Genomic Map