Assay Details
| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007568.5 | NP_031594.1 | 2 - 3 | 440 | 91 | |
| GenBank mRNA | AK046658.1 | - | 2 - 3 | 270 | 91 | |
| AK050206.1 | - | 2 - 3 | 256 | 91 | ||
| AK076272.1 | - | 2 - 3 | 360 | 91 | ||
| AK164633.1 | - | 2 - 3 | 441 | 91 | ||
| BC119152.1 | - | 2 - 3 | 306 | 8733901 | 91 | |
| BC119154.1 | - | 2 - 3 | 306 | 8733903 | 91 | |
| L08394.1 | - | 2 - 3 | 279 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Btc
Genomic Map